Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Timestrip™ PLUS Indicators
Record elapsed time at or above a specified temperature threshold in a simple and quick way
Supplier: MilliporeSigma™ Supelco™ 8082610EA
Description
- Small, self-adhesive labels that can be attached to or integrated into a product or device
- Manual activation button on top
- On window turns blue to confirm it is armed
- Blue color appears in the viewing window if a breach of the threshold temperature has occurred
- Cumulative duration of all breaches is shown by progress of blue dye along time markers
- Limited shelf life, use by expiration date on the label
- Time Accuracy: ±15% (under isothermal conditions)
- Recommended Storage: room temperature
- Produced under ISO 9001:2008
- Irreversible, single use
Specifications
48 Hr/8°C | |
48 Hour Run-Out Window (Calibrated (at 10°C)) | |
Recording Elapsed Time at or Above a Specified Temperature Threshold | |
8°C |
Aluminum/Plastic | |
40 x 19 mm | |
46°F | |
10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Timestrip Plus is a trademark of Timestrip Limited
Spot an opportunity for improvement?Share a Content Correction