Learn More
Electron Microscopy Sciences Thompson Monolayer Embedding Mold
Supplier: Electron Microscopy Sciences 70925
This mold used for embedding mammalian cells in monolayer culture in preparation of sectioning for light or electron microscopy. Tissue culture monolayer embedding molds eliminate the need for traditional BEEM capsules. The improved molds can be used to transfer the majority of cells from the entire surface of a cover slip to avoid losses of large amounts of cellular information occurring only randomly. In addition, other problems such as surface blemishes caused by air entrapment associated with BEEM capsules are eliminated.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.