Learn More
Hach Company THM Standard, 10 ppm as chloroform, 2 mL - pk/7
Supplier: Hach Company 2756707
For accuracy checks in Trihalomethane (THM) determinations. 10ppm as chloroform. 2-mL ampules - pack of 7