Learn More
New England Biolabs, Inc. Therminator™ DNA Polymerase – 200 units
SDP
Supplier:  New England Biolabs, Inc. M0261S
Encompass
Therminator DNA Polymerase is a 9N DNA Polymerase variant with an enhanced ability to incorporate modified substrates such as dideoxynucleotides, ribonucleotides and acyclonucleotides.
Catalog No. 50-811-685
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.