Learn More
Therapak™ Single-Specimen Collection
Suitable for on-site testing or remote site collection
Supplier: Therapak™ 40009
Description
- Specifically designed for substance abuse specimen collection
- Individually bagged
- Wide-mouth 6.5 oz. collection cup (without cap) is gender friendly for direct void; spout allows easy pouring of sample into transport vials
- Sterile, wide-mouth 90mL polypropylene containers are leak-resistant and feature an attached cap and sterility seal
- Temperature strip indicates sample temperature in °C and °F to ensure specimen integrity
- Available with transport bag constructed with two sealable tamper-evident pouches and absorbent material inside the specimen pouch
Specifications
6.5 oz. | |
192.2mL | |
For drugs of abuse testing, on-site testing or remote site collection |
Urine Specimen Collection Cup | |
Temperature strip | |
6.5 oz., 192.2mL |