Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylenebis(triphenylphosphonium bromide), 98+%, Thermo Scientific Chemicals

Catalog No. AAA1421418
Click to view available options
Quantity:
10 g
50 g

Tetramethylenebis(triphenylphosphonium bromide) is used in the synthesis of squalene. It is also used in the conversion of ?-diketones to cyclohexa-1,3-dienes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylenebis(triphenylphosphonium bromide) is used in the synthesis of squalene. It is also used in the conversion of ɑ-diketones to cyclohexa-1,3-dienes.

Notes
Hygroscopic. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 15546-42-6
Molecular Formula C40H38Br2P2
Molecular Weight (g/mol) 740.5
MDL Number MFCD00031714
InChI Key WTFXVDWCEPLDDY-UHFFFAOYSA-L
Synonym tetramethylenebis triphenylphosphonium bromide, 1,4-bis triphenylphosphonio butane dibromide, butane-1,4-diylbis triphenylphosphonium bromide, triphenyl 4-triphenylphosphaniumyl butyl phosphanium dibromide, tetramethylenebis triphenylphosphonium dibromide, triphenyl 4-triphenylphosphaniumylbutyl phosphanium dibromide, phosphonium,1,1'-1,4-butanediyl bis 1,1,1-triphenyl-, bromide 1:2, phosphonium,1,1'-1,4-butanediyl bis 1,1,1-triphenyl-,bromide 1:2
PubChem CID 3084237
IUPAC Name triphenyl(4-triphenylphosphaniumylbutyl)phosphanium;dibromide
SMILES C1=CC=C(C=C1)[P+](CCCC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6.[Br-].[Br-]

Specifications

Melting Point 292°C to 293°C
Quantity 50 g
Beilstein 3872522
Sensitivity Hygroscopic
Formula Weight 740.51
Percent Purity ≥98%
Chemical Name or Material Tetramethylenebis(triphenylphosphonium bromide)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-598-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.