Learn More
Neuromics, Inc Tet-Free Fetal Bovine Serum, 500 ml has undetectable levels of Tetracycline, making it an excellent option for research applications requiring regulated Tretracyclin levels.
Supplier:  Neuromics, Inc FBS002T
Encompass
Our FBS is manufactured in a fully integrated environment from raw material collection to finished product. It is produced to the highest standards in order to reduce risk and product variability, and to ensure product safety and performance.Our Tet-Free FBS features: Undetectable level of Tetracycline, Endotoxin levels : ≤10 EU/mL, Triple 0.1 μm Filtered, and Free of Virus and Mycoplasma.Tet-Free FBS removes Tetracycline. Excellent for research applications where Tetracycline must be regulated (for gene expression, etc.).
Catalog No. NC2071922
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.