Learn More
Enzo Life Sciences Testosterone high sensitivity ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901176
Encompass_Preferred
A highly sensitive competitive immunoassay for the measurement of Testosterone in human serum and plasma samples. Results are read on a colorimetric plate reader at 405 nm. Assay Time: 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-200-6898
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.