Learn More
Hach Company pH Test Strip, 7.5-14 pH units, 100 tests
Supplier: Hach Company 2751000
Most natural waters range from pH 4 to pH 9, but commonly are above pH 7 because of carbonates and bicarbonates (alkalinity). The pH of water used in industry, boiler feed water, and swimming pools usually is kept within a narrow range.