Learn More
Hach Company Hardness (Total) Test Kit, Model HA-DT
Supplier: Hach Company 2063600
Utilizing the Hach Digital Titrator, the Model HA-DT kit is used to determine hardness concentrations. Kit includes Digital Titrator, reagents and labware, manual and carrying case. Range: 10-4,000 mg/L. Contains approximately 100 tests.