Learn More
Hach Company Chlorine Dioxide Color Disc Test Kit, Model CLD-2
Supplier: Hach Company 2842800
Monitor ClO2 levels on-site with the Chlorine Dioxide Test Kit. The Chlorine Dioxide Test Kit uses the reliable DPD method to monitor ClO2 concentration, while eliminating chlorine interference through the addition of glycine. With 100 tests per kit and available range of 0-2 mg/L and resolution of 0.1 mg/L, operators can accurately monitor around the 0.8 mg/L regulated limit.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.