Learn More
Hach Company Alkalinity Test Kit, Model AL-DT
Supplier: Hach Company 2063700
Caused by the presence of carbonates, bicarbonates, hydroxides, and other dissolved salts, alkalinity is important to food and beverage processors, boiler operators, and aquatic biologists. Determine phenolphthalein and total alkalinity by visual endpoint titration. Using the Digital Titrator, this contains 100 tests with a range of 10-4,000 mg/L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.