Learn More
Chemglass Life Sciences Dual Temperature Controller, J-Kem, Apollo, Type ""T"" (-200C to 250C), Complete
SDP
Supplier:  Chemglass Life Sciences CG321001
Encompass_Preferred
Dual Temperature Controller, J-Kem, Apollo, Type "T" (-200C to 250C), Complete. J-Kem Scientific Apollo dual temperature controller features two independent temperature controllers in a single cabinet. Each independent channel has its own high power outlet, 100 hour timer to turn heating/cooling on or off at a user set time, and solid state over temperature protection circuit. Temperature ramping and serial communications are also provided with this unit.PLEASE NOTE: THE TYPE T, J AND K CONTROLLERS ARE SUPPLIED COMPLETE WITH: 1/8 DIAMETER X 12 LONG PTFE COATED THERMOCOUPLE, 10FT COILED EXTENSION CORD, AND A 14/20 PTFE THERMOCOUPLE ADAPTER. THE RTD UNIT IS THE CONTROLLER ONLY.Specifications:Dimensions: 4" H x 8" W x 9" DPower: 10 Amps @ 120VAC, 1200 Watts MAX/Channel, 1800 Watts (Total)
Catalog No. 50-401-833
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.