Learn More
Saf T Pak Inc Sepio Ambient Insulated Shipping System, 45-13L, 15° to 25°C
Supplier: Saf T Pak Inc 20623
The cost-effective SEPIO is a tested system offering a low barrier of entry when shipping payloads needing refrigeration, and can be used as an Overpack when transporting Category A Infectious Substances (UN 2814 or UN 2900).FEATURES: Key Features | Claims Pre-qualified for 2°C to 8°C and 15°C to 25°C using the same brick cassettesInmark Legacy ProfilesSeasonal and; Universal pack-outsCorrugated Cassettes for Easy HandlingMultiple payload sizesCost-effective solutionsWater-based refrigerant cassettesGlobal availability Temp/Profile/Duration:2-8/S120/W12015-25/S96/W72(+) Dead Weight (Includes Refrigerant)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.