Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ TeenPrep™ Lysing Matrix J
TeenPrep™ Lysing Matrix J, 15 mL Tube contains 2 mm yellow zirconia beads and 1.6 mm aluminum oxide particles. Will not bind nucleic acids.
Supplier: MP Biomedicals™ 116936005
Description
- Tube contains 2 mm yellow zirconia beads and 1.6 mm aluminum oxide particles.
- Used to grind animal and plant tissues, fixed samples and lyse fungi.
- For use on FastPrep™ instruments or any bead beater.
Certifications
Certified RNase and DNase free.
Specifications
15 mL | |
DNase Free | |
Sample Preparation, Sample lysis, Nucleic Acid isolation and purification, Molecular biology, Gene expression, Microbiological analysis, Gene function, Gene regulation | |
5 x 15 mL | |
Up to 2 g per prep |
Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Fixed Samples, Coral Emulsions, Mold, Animal Tissues, Fungi, Plants, Fixed Samples, Coral Emulsions, Mold | |
FastPrep-24, FastPrep 120; FastPrep-96 | |
Lysing Matrix J | |
RNase Free |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction