Learn More
New England Biolabs, Inc. Taq 2X Master Mix – 500 reactions (50 µl vol)
SDP
Supplier:  New England Biolabs, Inc. M0270L
Encompass
Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5' to 3' polymerase activity and a 5' flap endonuclease activity. Taq 2X Master Mix is an optimized ready-to-use solution containing Taq DNA Polymerase, dNTPs, MgCl2, KCI and stabilizers. It is ideally suited to routine PCR applications from templates including pure DNA solutions, bacterial colonies, and cDNA products. It can amplify up to 4 kb from complex genomic DNA or up to 5 kb from lambda DNA.
Catalog No. 50-995-170
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.