Learn More
Biotang Inc TAE (50X PH 8.0) 1000ML. BM-250. 2 M Tris-base, 1M acetate and 50 mM EDTA, pH 8.0 +/- 0.2. Prepared in 18.2 megohms water and filtered through 1 micron filter.
Supplier: Biotang Inc BTBB091
TAE (50X PH 8.0) 1000ML. BM-250. 2 M Tris-base, 1M acetate and 50 mM EDTA, pH 8.0 +/- 0.2. Prepared in 18.2 megohms water and filtered through 1 micron filter.