Learn More
Cole-Parmer Electro Numerics PLT-5000 Contact/Noncontact Laser Pocket Tachometer
Supplier: Cole-Parmer 8760020
This pocket tachometer measures both contact and noncontact readings. Laser allows noncontact measurement up to 60" (152.4 cm) away. Select from 10 measurements including min/max and final. Also features 14 memory points, overrange indicator, one-second display update, and low-battery indicator.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.