Learn More
Cole-Parmer Extech RPM10 Combination Laser Tachometer with Infrared Temperature
Supplier: Cole-Parmer 8770220
This tachometer measures contact rpm and surface speed, noncontact rpm via laser, and noncontact temperature with infrared capabilities. For maximum safety, measure from up to 6 ½ (1.9 cm) feet away from the target. Noncontact temperature offers quick and safe surface temperatures for high temperature applications or in hard-to-reach areas. Other features include last reading, min and max memory, and display flips according to measurement mode.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.