Learn More
New England Biolabs, Inc. T4 Gene 32 Protein - 100 ug
SDP
Supplier:  New England Biolabs, Inc. M0300S
Encompass
T4 Gene 32 Protein - 100 ug. T4 Gene 32 Protein is a single-stranded DNA (ssDNA) binding protein required for bacteriophage T4 replication and repair. It cooperatively binds to and stablizes transiently formed regions of ssDNA and plays an important structural role during T4 phage replication. It also has been used extensively to stabilize and mark regions of ssDNA for electron microscopic examination of intracellular DNA structures. Recently, it has been shown to improve restriction enzyme digestion, improve the yield and efficiency of reverse transcription (RT) reactions during RT-PCR, enhance T4 DNA polymerase activity, as well as increase the yield of PCR products.Help NEB go green. FREE FREIGHT on NEB orders greater than $350.00!
Catalog No. 50-994-960
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.