Learn More
Andwin Scientific 1/2" Tough-Spots 1,000/roll, Blue
SDP
Supplier:  Andwin Scientific TSPOTS50B
Encompass_Preferred
Circular cryogenic labels formatted for handwriting. Tough Spots on a roll are easy to write PVC cryogenic labels that accept writing from most lab markers. Tough Spots come in a variety of colors and sizes to ensure the right label for the right job!
Catalog No. NC1663570
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.