Learn More
Creative Biomart Recombinant T.pallidum Partial p17 Antigen, His-tagged
Supplier:  Creative Biomart TPP430
Encompass_Preferred
The E. coli derived recombinant protein is fused at N-terminus with 6 His tag and contains the Trp. Pallidum p17 immunodominant regions.
Catalog No. 50-208-4808
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.