Learn More
Enzo Life Sciences T-2 Toxin (1mg). CAS: 21259-20-1
SDP
Supplier:  Enzo Life Sciences ALX630101M001
Encompass_Preferred
Mycotoxin. Induces DNA damage and apoptosis. Increases blood-brain barrier permeability and inhibits monoamine oxidase activity in the brain. Alternative name: 4beta,15-Diacetoxy-3alpha-hydroxy-8alpha-(3-methylbutyryloxy)-12,13-epoxytrichothec-9-ene. Purity: ≥98% (TLC). Appearance: White to off-white crystalline solid. Solubility: Soluble in methylene chloride (5mg/ml) or chloroform. Long Term Storage: -20°C.
Catalog No. 50-200-9623
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.