Learn More
Biotang Inc E-Z Syringe tips 2.5ML STERILE, 100/PK
Supplier:  Biotang Inc 702389
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Fit most manufacture's repeating pipettes using typical standard sized syringe tips, doesn't fit Eppendorf models. Made in Germany.
Catalog No. 50-751-5005