Learn More
Fisherbrand™ Synthetic-Tipped Applicators
Polyester-tipped applicators on wooden or plastic handles
Supplier:  Fisherbrand™ 22363165
 View more versions of this product
Catalog No. 22-363-165
Description
Description
  • Multi-purpose applicators ideal for specimen collection and use in a wide range of diagnostic testing and screening
  • Sterile and non-sterile products available (see product specifications)
  • Polyester-tipped and cotton applicators on wooden or plastic handles available (see product specifications)
  • Swabs are not certified DNA, DNase or RNase free
  • Not suitable for Forensic use or Human Genetic testing
Specifications
Specifications
Wood
6 in.
Bulk
Specimen collection and transport system
Polyester
15 cm
Non-sterile
Applicator
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.