Learn More
Med Vet International Ethicon Vicryl Suture, Size 5-0, PS-2, 18", 12/Box
Supplier:  Med Vet International J495G
Encompass_Preferred
The Ethicon J495G suture is a high-quality absorbable Polyglactin 91 surgical suture. It measures 18in. in length, has a size of 5-, they are undyed and typically have an absorption rate of 56-7 days. This suture comes with a PS-2 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-209-2819
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.