Learn More
Med Vet International Silk Suture Cassette, Size 2, 100M Veterinary Use Only, each
Supplier: Med Vet International SLK2
Non-absorbable silk braided sutureExtremely strong tensile strength. High tensile strength suture that exceeds USP requirements. Excellent knot securityDesigned for secure knotting. Smooth surface allows for easy passage through tissue. Composition: Silk. Absorption Type: Non-Absorbable. Structure: Braided. Thread Color: Black
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.