Learn More
Med Vet International Silk Suture Cassette, Size 2-0, 100M Veterinary Use Only, each
Supplier:  Med Vet International SLK20
Encompass_Preferred
Non-absorbable silk braided sutureExtremely strong tensile strength. High tensile strength suture that exceeds USP requirementsExcellent knot securityDesigned for secure knottingSmooth surface allows for easy passage through tissue. Composition: Silk. Absorption Type: Non-Absorbable. Structure: Braided. Thread Color: Black
Catalog No. 50-264-3438
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.