Learn More
Med Vet International Oasis PDO Suture Cassette, Size 1, Length of 15M, Each
Supplier:  Med Vet International PD01
Encompass_Preferred
The veterinary use Oasis PDO suture cassette is a monofilament synthetic absorbable suture. The PDO smooth monofilament suture has a gradual absorption rate between 18 and 22 days. It retains 5% of its tensile strength for up to 6 weeks. This suture provides excellent pliability and holding strength. Size: 1. Length: 15M.
Catalog No. 50-264-3464
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.