Learn More
Med Vet International Ethicon Chromic Gut Suture, Size 5-0, P-3, 18", 12/Box, 687G
Supplier:  Med Vet International 687G
Encompass_Preferred
The Ethicon 687G suture is a high-quality absorbable Chromic Gut surgical suture. It measures 18 in. length, size of 5-0, dyed brown to provide greater visibility while suturing, and typically have an absorption rate of 90 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. NC1656446
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.