Learn More
Chemglass Life Sciences Support Jack, Anodized Aluminum, 15.7" x 15.7" Deck, 18.5" Max Height, 176lbs Max Load
SDP
Supplier:  Chemglass Life Sciences CG305925
Encompass_Preferred
Support Jack, Anodized Aluminum, 15.7" x 15.7" Deck, 18.5" Max Height, 176lbs Max Load. Support Jack deck is constructed of either Stainless Steel or Anodized Aluminum, while the lift mechanism for both is constructed entirely from Type 304 stainless steel and PTFE. This design provides long life, especially when stored and used in corrosive environments. Support Jack is resistant to sanitizing agents and autoclavable for sterile applications. Jack can be used to support flasks, jars, columns, large baths, magnetic stirrers, heaters, and more. They are popular for use in a wide variety of environments, including fume hoods, solution-baths, and marine and industrial labs. Optional ratchet wrench is available for larger sizes. Two-year warranty on all Support Jacks.
Catalog No. CG305925
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.