Learn More
Cytiva (Formerly Pall Lab) Supor 450 Membrane Disc Filters, 0.45 m - 142 mm, no tab (25/pkg)
Supplier:  Cytiva 66553
Encompass_Preferred
High Flow Rate Membrane Optimized for Biological, Pharmaceutical, and Sterilizing Filtration Requirements-Fast filtration with superior flow rates and high throughputs -Low protein binding and extensive drug compatibility for critical applications-Saves time and money with fewer filter changes per sample volume-142 and 293 mm discs feature printed tab for instant recognition of pore size and lot number This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3334
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.