Learn More
MilliporeSigma™ Supelco™ Super Clean™ (Base-Plate Design) Gas Purifiers
For the Purification and Management of GC Gas Streams
Supplier: MilliporeSigma™ Supelco™ SU861021
Description
- Provides gas at the proper purity level and pressure, based on its intended use
- The purifiers connect into a base plate in such a manner that gas supply is not interrupted during purifier change-out
- Purifier can be replaced without shutting down the entire system
- Eliminates the risk of forcing a slug of air into the instrument after purifier change-out
Specifications
Enhancing The Performance of GC Detectors | |
1 Ea. | |
Moisture |
GC Detectors | |
With Indicator |