Learn More
Electron Microscopy Sciences Sudan Black, Certified, C.I. DcZb-2 25 GR
Supplier: Electron Microscopy Sciences 21610
(Solvent Black 3)
CAS #4197-25-5 C₂9H₂4N₆ F.W. 456.54
Solubility: Water-insoluble; Alcohol-soluble; 4.0% Cellosolve; 1.0% Glycol
Solvent Black 3 is a stain for fat in animal tissue and as a tain for chromosomes, golgi, and leucocyte granules. C29H24N6 M.W. 456.56
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.