Learn More
Sigma Aldrich Fine Chemicals Biosciences Suba-Seal(R) septa white rubber, Suba Seal, 33, fits neck I.D., 17.5 mm, for use with all Sure/Seal bottles | 100EA
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z167290100EA
Suba-Seal(R) septa white rubber, Suba Seal, 33, fits neck I.D., 17.5 mm, for use with all Sure/Seal bottles | 100EA