Learn More
World Precision Instrument SU-MPC385 Motorized Micromanipulator Systems-505088
SDP
Supplier:  World Precision Instrument 505088
Encompass_Preferred
  • Quietest electronics in the industry optimized for single channel recording
  • Single controller and ROE will run two stepper motor drive manipulators
  • Self-detecting, daisy-chain capability allows control of four manipulators from one 505093
  • User-friendly interface: single button access to all major functions
  • Easily configurable virtual 4th axis set without external computer
  • Accelerated Mode for fast, manual manipulator movement
  • Easy toggle selection of Mode (speed/resolution, pulsed diagonal, Accelerated Mode)
  • Display indicates X, Y, Z coordinates, Mode, active manipulator
  • Robotic Home and Work Position moves for easy automated pipette exchange
  • Faster robotic moves than previous versions
  • Definable 4th axis for coaxial pipette movement, angle selected by DIP switches
  • Simple USB interface
Catalog No. 50-253-9255
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.