Learn More
Research Products International Corp Styrofoam Micro-Tube Storage Boxes, 2 3/4 High Inches,10 per Case
SDP
Supplier:  Research Products International Corp 145746
Encompass_Preferred
Styrofoam storage boxes offer a safe and efficient method for handling a variety of biological samples: freezing, storing, transporting. Storage boxes accommodate up to 100 micro-tubes (1.5 and 2.0 ml) in a 5 x 20 array.
Catalog No. 50-550-191
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.