Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium Standard, 1000μg/mL in water with dilute nitric acid, Ultra Scientific

Catalog No. USICC108
Encompass
Click to view available options
Quantity:
125 mL

Chemical Identifiers

CAS 7440-24-6, 7697-37-2, 7732-18-5
Molecular Formula Sr
Molecular Weight (g/mol) 87.62
InChI Key CIOAGBVUUVVLOB-UHFFFAOYSA-N
PubChem CID 5359327
ChEBI CHEBI:33324
IUPAC Name strontium
SMILES [Sr]

Specifications

Quantity 125 mL
CAS Max % 99.7, 0.2, 0.1
Concentration or Composition (by Analyte or Components) 1000μg/mL
Analyte or Component Names Water,Nitric Acid,Strontium
Solvent or Matrix Water with dilute nitric acid
Chemical Name or Material Strontium Standard
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.