Learn More
MSC Made in USA 6206-010 9 Inch Long x 3.6614 Inch Wide, Stroboscope 3.5433 Inch Meter Thickness, 30 to 10,000 Flash per Minute
Supplier:  MSC 60100567
Encompass
Made in USA 6206-010 9 Inch Long x 3.6614 Inch Wide, Stroboscope 3.5433 Inch Meter Thickness, 30 to 10,000 Flash per Minute
Catalog No. NC1904623
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.