Learn More
Vector Laboratories Streptavidin Horseradish Peroxidase Concentrate, For IHC 1 mg
SDP
Supplier:  Vector Laboratories SA5004
Encompass_Preferred
Vector Laboratories streptavidin and avidin reagents are highly purified and possess very low non-specific binding properties. They have a high affinity for biotin and can be used to detect biotinylated secondary antibodies and other macromolecules in applications such as immunohistochemistry and in situ hybridization. Chromogenic signals can be amplified using biotinylated secondary antibodies followed by HRP-labeled streptavidin or avidin. Horseradish Peroxidase Streptavidin (HRP Streptavidin) is produced by a coupling procedure that preserves the high specific activity of the peroxidase. Using biotinylated primary or secondary intermediates and peroxidase-labeled streptavidin, antigens can be localized in histological sections, cytospin preparations, or smears.
Catalog No. NC9499525
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.