Learn More
MTC Bio Strainer Cap for FlowTubes, 35µm Strainer Mesh, Sterile, 25 per Bag, Pack of 500
Supplier: MTC Bio T9009
Strainer cap contains 35µm nylon mesh screen. Assists in cell sorting/straining applications. For use with FlowTubes 12x75mm culture tubes. Supplied in pack of 500 (20 bags of 25 caps).