Learn More
Electron Microscopy Sciences Straight, Fine Point Diamond Scriber
Supplier: Electron Microscopy Sciences 62107ST
This diamond scriber is the ultimate scribing instrument offering the most precision for the most delicate applications. It is ideal for fine scribing on silicon wafers and glass coverslips under the microscope and for precision scribing and repairing thin film circuits and microcircuits.
The diamond mountings are 0.8 mm diameter with a reduced shank size of 0.5 mm or 0.25 mm diameter, which holds the diamond stylus. The natural diamond has a 60 degree included angle and a 0.12 mm tip radius. The integrated 6.35 mm anodized hexagonal (non-rolling) aluminum handle has a positive finger grip in the form of grooves for easy control and nonslippage. Packaged in a protective plastic tube.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.