Learn More
Saf T Pak Inc STP 309KIT CAT B ACCESSORIES
SDP
Supplier:  Saf T Pak Inc 35131
Encompass_Preferred
Saf-T-Pak® STP 309KIT Cat B Accessories 8 case STP-309/308System Components:Absorbent Sheet 4x6 (8 ea.)Saf-T-Rap® Sticky Bubblewrap, 6 x 12” (8 ea.)STP-710 Tyvek Envelope (Medium) (8 ea.)STP-711 Inner Polybag (Medium) (8 ea.)STP-804 Dry Ice Label (8 ea.)STP-819 Exempt Human/Animal Specimen Marking (8 ea.)Class 9 Hazard Label (8 ea.)
Catalog No. NC2219252
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.