Learn More
Stone Medical Clean Collect™ Blood Culture Kits
Blood culture diversion system prevents contamination in peripheral blood cultures
Supplier:  Stone Medical CC225R
 View more versions of this product
Catalog No. 23-200-106
Description
Description
Reducing blood culture contamination rates prevents unnecessary treatment of patients. Clean Collect kits can help to increase quality and save considerable resources.
  • Prevents blood culture contamination using proprietary diversion method
  • Average contamination rate of 0.5% compared to 3% national average
  • Diversion method reduces contamination rates by an average of 74%
  • Standardized kits promote consistency for long term improvement
  • Reduces hospital costs considerably
  • Ambient temperature (0-40°C) dry storage; 10 month shelf life
Specifications
Specifications
Blood Culture Kit
Diversion tube, BD push button butterfly, blood culture bottle adaptor, Prevantics Swabstick antiseptic, sterile gloves, tourniquet, alcohol pads, bandage, instruction card, tracking decals
Ambient temperature (0-40°C) dry storage
Diversion tube, BD push button butterfly, blood culture bottle adaptor, Prevantics™ Swabstick antiseptic, sterile gloves, tourniquet, alcohol pads, bandage, instruction card, tracking decals
Compatible with BacT/Alert Systems
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.