Learn More
STEMCELL Technologies STEMscript™ cDNA Synthesis Kit with Oligo(dT) Primers, Size: 1 Kit
SDP
Supplier:  STEMCELL Technologies 79003
Encompass_Preferred
STEMscript™ cDNA Synthesis Kit includes three components that have been optimized to provide efficient synthesis of first-strand cDNA in preparation for qPCR amplification: STEMscript™ Reverse Transcriptase Mix (contains STEMscript™ Reverse Transcriptase purified from recombinant E. coli, and recombinant ribonuclease inhibitor), STEMscript™ Reaction Buffer (contains primers, MgCl₂, and dNTPs), and nuclease-free water. This kit can be used to reverse transcribe total RNA, mRNA, or synthetic transcript RNA, and is required for gene expression analyses such as qPCR and RNA sequencing. STEMscript™ cDNA Synthesis Kit is available with either oligo(dT) or random primers. Oligo(dT) primers anneal to the poly(A) tails of eukaryotic mRNAs, whereas random primers are used for reverse transcription of RNA without poly(A) tails, such as rRNA, tRNA, degraded RNA, RNA isolated from prokaryotic sources, and RNA with known secondary structures.
Catalog No. 50-197-6788
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.