Learn More
Clontech Labs 3P Stellar™ Competent Cells, 10 x 100 ul
Supplier: Clontech Labs 3P 636763
Stellar Competent Cells are an E. coli HST08 strain that provides high transformation efficiency. These cells can be used in a wide variety of applications—from preparation of cDNA and genomic libraries, to construction of longer-length genomic libraries, to sub-cloning, and even methylated DNA cloning. Stellar Competent Cells lack the gene cluster for cutting foreign methylated DNA (mrr-hsdRMS-mcrBC and mcrA), and are therefore useful for cloning methylated DNA. The cells can also be used for blue/white screening (i.e., α-complementation) when transformed with vectors containing the lacZα gene. A pUC19 vector is provided as well as SOC Medium. Stellar Competent Cells are recommended for use with Clontech's In-Fusion PCR Cloning Kits. 10 x 100 ul
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.