Learn More
Starplex™ Scientific Histoplex™ Histology Containers
Specifically engineered for the storage of histology specimens
Supplier:  Starplex™ Scientific H50002C
 View more versions of this product
Catalog No. 15-182-81
Description
Description
Specifically engineered for the storage of histology specimens
  • O-ring polyethylene screw cap forms a seal that reduces most formalin leakage and evaporation
  • Durable polypropylene containers are translucent, flexible, resist cracking and splitting, and eliminate problems associated with formalin evaporation
  • 40mL to 1L containers have graduations for easy fill line viewing
  • 2 to 8L containers are white polypropylene with unique tamper-evident snap-on lids
Specifications
Specifications
Yes
Polypropylene
Clear
Clear
In case
5L
Non-sterile
Snap cap
Tamper evident pull
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.