Learn More
New England Biolabs, Inc. Standard Taq Reaction Buffer Pack – 6 ml (4 x 1.5 ml)
Supplier: New England Biolabs, Inc. B9014S
New England Biolabs supplies a unique 10X reaction buffer for use with Taq DNA Polymerase in reaction systems which require a detergent free, low ionic strength buffer.