Learn More
Accuflow Systems Stainless Steel Rack
Supplier:  Accuflow Systems HF252
Encompass_Preferred
Stainless Steel horizontal rack to hold 25 standard 2” freezer boxes
Catalog No. NC3136624
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.