Learn More
Hach Company Stablcal Turbidity Standard, 1.0 NTU, 100 mL
Supplier:  Hach Company 2659842
Encompass_Preferred
Stablcal Turbidity Standard for all Turbidimeters. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. NTU Value: 1.0. Bottle: 100 mL. For TL23, TU52, 2100Q, 2100N, 2100AN series.
Catalog No. 50-199-6480
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.